Proteinase K Powder, 500 mg - FAQs
Proteinase K Powder, 500 mg - FAQs
View additional product information for Proteinase K Powder - FAQs (AM2544, AM2542)
1 product FAQs found
How do I isolate DNA from single bacterial colonies before PCR?
Scrape a single colony from the surface of an agar plate and transfer to 12 µL of SCL solution [10 mM Tris-HCl (pH 7.5), 1 mM EDTA, 50 µg/mL proteinase K] and incubate for 15 min at 55 degrees C. Inactivate proteinase K for 15 min at 80 degrees C and add 20 µL of deionized water. Centrifuge at 12,000 x g for 3 min. Transfer the supernatant to a new tube.