SUPERase·In™ RNase Inhibitor (20 U/μL), 10,000 U - FAQs
SUPERase·In™ RNase Inhibitor (20 U/μL), 10,000 U - FAQs
View additional product information for SUPERase·In™ RNase Inhibitor (20 U/μL) - FAQs (AM2696, AM2694)
2 product FAQs found
What temperature and pH is SUPERase·In RNase Inhibitor (Cat. No. AM2694, AM2696) active up to?
SUPERase·In RNase Inhibitor (20 U/µL) (Cat. No. AM2694, AM2696) is active up to 65 degrees C and over a pH range of 5.5 to 8.5.
Find additional tips, troubleshooting help, and resources within our Nucleic Acid Purification and Analysis Support Center.
What kind of Rnases can be inhibited using SUPERase.In RNase Inhibitor?
SUPERase.In RNase Inhibitor is a protein-based inhibitor of nonhuman origin that noncovalently binds and inhibits the most common and troublesome RNases, including RNase A, B, C, 1, and T1.