T7 RNA Polymerase-Plus™ Enzyme Mix (200 U/μL) - FAQs
T7 RNA Polymerase-Plus™ Enzyme Mix (200 U/μL) - FAQs
View additional product information for T7 RNA Polymerase-Plus™ Enzyme Mix (200 U/μL) - FAQs (AM2718)
1 product FAQs found
Can I substitute the enzyme mix in the MEGAscript T7 Transcription Kit (Cat. No. AM1333) with T7 RNA Polymerase-Plus Enzyme Mix (Cat. No. AM2718)?
No, we do not recommend substituting the enzyme mix in the MEGAscript T7 Transcription Kit (Cat. No. AM1333) with T7 RNA Polymerase-Plus Enzyme Mix (Cat. No. AM2718).