Product Image
Invitrogen™

siPORT™ Amine Transfection Agent with manual

A polyamine-based Ambion™ siRNA transfection agent that can be used to transfect cells during subculturing-—saving a full day of valuableRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityProduct Type
AM4502400 μLTransfection Reagent
AM4503M1 mLTransfection Reagent with Manual
AM45031 mLTransfection Reagent
AM4502M400 μLTransfection Reagent with Manual
Catalog number AM4502
Price (USD)
860.00
Each
Quantity:
400 μL
Product Type:
Transfection Reagent
Price (USD)
860.00
Each
A polyamine-based Ambion™ siRNA transfection agent that can be used to transfect cells during subculturing-—saving a full day of valuable time—without increasing toxicity. It is supplied in one tube containing 1 mL. A successful siRNA experiment requires the appropriate transfection agent for your cells, and one that is optimized for siRNA delivery. The efficiency with which different transfection reagents deliver nucleic acids can vary dramatically with the type of nucleic acid being transfected (RNA vs. DNA, single-stranded vs. double-stranded) and the cells being used.

Accessory Products:
siPORT™ NeoFX™ Transfection Agent (SKU#s AM4511 and AM4510), a cationic and neutral lipid mixture, is also available. For testing which transfection agent is best with your system, the Silencer™ siRNA Transfection II Kit (SKU# AM1631) is recommended. It contains the two different transfection agents along with controls for transfection optimization.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
ClassificationAnimal Origin-Free
For Use With (Application)Transfection
High-throughput CompatibilityNot High-throughput Compatible (Manual)
Product LineAmbion, siPORT
Product TypeTransfection Reagent
Quantity400 μL
Serum CompatibleYes
Shipping ConditionWet Ice
Cell TypePrimary Cells
Format6-well Plate, 12-well Plate, 24-well Plate, 48-well Plate, 96-well Plate, Flasks
Sample TypeSynthetic siRNA
Transfection TechniqueLipid-based Transfection
Unit SizeEach
Contents & Storage
Store at 4°C.