RNA<i>later</i>&trade; Stabilization Solution
RNA<i>later</i>&trade; Stabilization Solution
RNA<i>later</i>&trade; Stabilization Solution
Invitrogen™

RNAlater™ Stabilization Solution

RNAlater RNA Stabilization Solution stabilizes and protects cellular RNA in intact, unfrozen tissue samples, eliminating the need to immediately process tissue samples or to freeze samples in liquid nitrogen for later processing.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
AM702250 x 1.5 mL
AM702320 x 5 mL
AM7020100 mL
AM70241 x 250 mL
AM7021500 mL
Catalog number AM7022
Price (USD)
342.65
Online exclusive
378.00
Save 35.35
Each
Quantity:
50 x 1.5 mL
Price (USD)
342.65
Online exclusive
378.00
Save 35.35
Each

RNAlater RNA Stabilization Solution stabilizes and protects cellular RNA in intact, unfrozen tissue samples, eliminating the need to immediately process tissue samples or to freeze samples in liquid nitrogen for later processing. Tissue pieces can be harvested and submerged in RNAlater RNA Stabilization Solution for storage without jeopardizing the quality or quantity of RNA obtained after subsequent RNA isolation. Advantages of using RNAlater RNA Stabilization Solution:

  • Effectiveness – stabilize RNA for 1 day at 37°C, 1 week at 25°C, 1 month at 4°C, or indefinitely at -20°C
  • Simplicity – a single reagent that immediately inactivates RNases and stabilizes RNA within tissues or cells
  • Convenience – no need to freeze samples in liquid nitrogen or rush samples back to the lab freezer
  • Mobility – perfect for tissue collection 'in the field'
  • Versatility – compatible with many RNA isolation procedures, including most RNA isolation kits

Applications
RNAlater RNA Stabilization Solution has been tested on a variety of mammalian tissues, plants, E. coli, Xenopus, fish, and Drosophila. It is ideal for:

  • Protecting RNA integrity in tissues rich in RNases
  • Collecting samples from different time points without having to process the samples from each time point immediately
  • Archiving tissues for future microdissection
  • Submerging animal cavities or organs to stabilize RNA during lengthy dissection procedures
  • Collecting samples at locations (e.g., hospitals, field sites, the space shuttle) where immediate RNA isolation is not possible
  • Shipping samples on wet ice or even at room temperature if shipped overnight

RNAlater RNA Stabilization Solution procedure
The dissected tissue (less than 0.5 cm in any one dimension) is simply submerged in approximately 5 volumes of RNAlater solution at room temperature. The solution permeates the cells, stabilizing the RNA. The sample can then be stored indefinitely at -20°C (the tissue does not freeze), at 4°C for up to a month, or at 25°C for up to a week. For RNA isolation, the tissue is simply removed from RNAlater solution and treated as though it had just been harvested. Most tissues can be transferred directly to a lysis buffer and homogenized. Samples treated with RNAlater solution and then frozen can be ground with mortar and pestle or thawed and processed like fresh tissue without concern for cell rupture and release of RNases since the RNases have already been inactivated. Cells can be spun out and then added to lysis buffer, or in some cases, RNAlater solution can be added along with the cells directly to the lysis buffer.

Compatible with a variety of procedures
RNAlater RNA Stabilization Solution is compatible with one-step RNA isolation methods, such as TRIzol Reagent; with glass binding methods such as Qiagen's RNeasy or the Ambion RNAqueous kit; with acid phenol extraction methods such as the Ambion ToTALLY RNA kit; and with methods that use oligo(dT) selection of mRNA, such as the Ambion Poly(A)Purist™ kit. In-house research, as well recently published independent research, indicates that the use of RNAlater RNA Stabilization Solution for tissue storage does not affect the outcome of subsequent RNA expression analysis experiments compared to other processing methods.

For Research Use Only. Not for use in diagnostic procedures.

Specifications
DescriptionRNAlater™ Stabilization Solution
FormLiquid
Product LineAmbion, RNAlater
Product TypeRNA Storage Reagent
Quantity50 x 1.5 mL
Shipping ConditionRoom Temperature
Unit SizeEach
Contents & Storage
Store at room temperature.

Frequently asked questions (FAQs)

I see a precipitate in my RNAlater Stabilization Solution. What should I do?

Heat the solution to 37 degrees C for 15 minutes and agitate to redissolve it.

Find additional tips, troubleshooting help, and resources within our RNA Sample Collection, Protection, and Isolation Support Center.

What can I use to protect RNA in my frozen tissue sample?

RNAlater-ICE Frozen Tissue Transition Solution can be used to submerge a frozen sample, then thaw it overnight at -20 degrees C or colder. Once thawed, tissues can then be processed like fresh tissues using standard RNA isolation procedures.

Find additional tips, troubleshooting help, and resources within our RNA Sample Collection, Protection, and Isolation Support Center.

Can RNAlater Stabilization Solution be used to preserve samples for laser capture microdissection (LCM)?

While we have not tested this in-house, there is evidence that it is possible to use tissues preserved in RNAlater Stabilization Solution and then perform laser capture microdissection. Please see the reference below:
Thelen P, Burfeind P, Grzmil M et al. (2004) cDNA microarray analysis with amplified RNA after isolation of intact cellular RNA from neoplastic and non-neoplastic prostate tissue separated by laser microdissections. Int J Oncol 24(5):1085-1092.

Find additional tips, troubleshooting help, and resources within our RNA Sample Collection, Protection, and Isolation Support Center.

Have you tested temperature stability of RNA stored in RNAlater Stabilization Solution?

RNA stored in RNAlater Stabilization Solution is stable for 1 day at 37 degrees C, 1 week at 25 degrees C, 1 month at 4 degrees C, or long-term at -20 degrees C.

Find additional tips, troubleshooting help, and resources within our RNA Sample Collection, Protection, and Isolation Support Center.

Can cells treated with RNAlater Stabilization Solution be FACS sorted? How about mass spectrometry?

Yes, you should be able to sort cells by fluorescence-activated cell sorting (FACS) after treatment with RNAlater Stabilization Solution. If you run into problems due to the viscosity of RNAlater Stabilization Solution, you may need to dilute it with cold nuclease-free water. This will not affect the protection of the RNA. Samples should be processed quickly and as cold as possible. Read more about this procedure and view references here (http://www.thermofisher.com/us/en/home/references/Invitrogen-tech-support/rna-isolation/tech-notes/facs-into-rnalater-solution-for-gene-profiling.html). You should also be able to perform mass spectrophotometry on samples treated with RNAlater Stabilization Solution. Please note that salt can form adducts on the protein at the desorption/ionization step. To avoid this problem, dialyze the sample to get rid of all salt.

Find additional tips, troubleshooting help, and resources within our RNA Sample Collection, Protection, and Isolation Support Center.

Citations & References (1)

Citations & References
Abstract
20 beta-hydroxysteroid dehydrogenase and CYP19A1 are differentially expressed during maturation in Atlantic cod (Gadus morhua).
Authors:Mittelholzer C, Andersson E, Consten D, Hirai T, Nagahama Y, Norberg B,
Journal:J Mol Endocrinol
PubMed ID:17909270
In order to better quantify the molecular mechanisms regulating final oocyte maturation and spawning, complete coding sequences with partially or fully untranslated regions for the steroidogenic enzymes, cytochrome P450 aromatase and 20 beta-hydroxysteroid dehydrogenase, were cloned from ovaries of Atlantic cod (Gadus morhua). The nucleotide and amino acid sequences showed ... More