RNA Control 250 with Manual, 2 x 20 μL, with Manual - FAQs
RNA Control 250 with Manual, 2 x 20 μL, with Manual - FAQs
View additional product information for RNA Control 250 with Manual - FAQs (AM7155, AM7155M)
1 product FAQs found
What is the sequence of RNA Control 250?
The sequence of RNA Control 250 is proprietary.