Human Brain Total RNA - FAQs
Human Brain Total RNA - FAQs
View additional product information for Human Brain Total RNA - FAQs (AM7962)
2 product FAQs found
Do you offer isolated, purified RNA?
Yes, please visit this website (http://www.thermofisher.com/us/en/home/life-science/dna-rna-purification-analysis/rna-extraction/rna-types/purified-rna.html) for our FirstChoice total RNA isolated from human tissues, as well as from E. coli and human cell lines.
Does Human Brain Total RNA (Cat. No. AM7962) come from pooled donors or a single individual? Where can I find the demographic information for the product I receive?
The Human Brain Total RNA (Cat. No. AM7962) is isolated from one individual donor. The donor demographics, such as race, gender, age, and cause of death (COD), can be found on the certificate of analysis (COA).
You can find the COA for a specific lot number by using the Documents and Certificatessearch page or by filling out a Certificate of Analysis (COA) or Certificate of Origin (COO) Request Form.
Find additional tips, troubleshooting help, and resources within our RNA Sample Collection, Protection, and Isolation Support Center.