Human Kidney Total RNA - FAQs
Human Kidney Total RNA - FAQs
View additional product information for Human Kidney Total RNA - FAQs (AM7976)
1 product FAQs found
Do you offer isolated, purified RNA?
Yes, please visit this website (http://www.thermofisher.com/us/en/home/life-science/dna-rna-purification-analysis/rna-extraction/rna-types/purified-rna.html) for our FirstChoice total RNA isolated from human tissues, as well as from E. coli and human cell lines.