MagMAX™ Wash Solution 1 Concentrate - FAQs
MagMAX™ Wash Solution 1 Concentrate - FAQs
View additional product information for MagMAX™ Wash Solution 1 Concentrate - FAQs (AM8504)
1 product FAQs found
How should I prepare MagMAX wash solutions 1 and 2 from standalone Wash Solution 1 Concentrate (Cat. No. AM8504) and Wash Solution 2 Concentrate (Cat. No. AM8640)?
For MagMAX Wash Solution 1 Concentrate (Cat. No. AM8504), add 70 mL isopropanol to 135 mL of wash buffer concentrate. This can be added directly to the AM8504 bottle for a total volume of 205 mL of wash solution 1. For MagMAX Wash Solution 2 Concentrate (Cat. No. AM8640), add 160 mL ethanol to 40 mL of wash buffer concentrate. This can be added directly to the AM8640 bottle for a total volume of 200 mL of wash solution 2.