Product Image
Invitrogen™

Gel Loading Solution (All-Purpose, Native Agarose)

A 10X solution of 40% Sucrose, 0.17% Xylene Cyanol, and 0.17% Bromophenol Blue. Supplied in one 1.4 mL tube. All자세히 알아보기
Have Questions?
카탈로그 번호수량
AM85561 mL
카탈로그 번호 AM8556
제품 가격(KRW)
102,000
온라인 행사
Ends: 30-Sep-2026
116,000
할인액 14,000 (12%)
Each
수량:
1 mL
대량 주문 또는 맞춤형 요청
제품 가격(KRW)
102,000
온라인 행사
Ends: 30-Sep-2026
116,000
할인액 14,000 (12%)
Each
A 10X solution of 40% Sucrose, 0.17% Xylene Cyanol, and 0.17% Bromophenol Blue. Supplied in one 1.4 mL tube. All Ambion® Gel Loading Solutions are rigorously tested for non-specific endonuclease activity, exonuclease activity, RNase activity, and for functionality. These are the same solutions we use in-house and in our kits.
For Research Use Only. Not for use in diagnostic procedures.
사양
버퍼Sample Loading Buffers
농도10X
젤 호환성Polyacrylamide Gels, Native Agarose
수량1 mL
배송 조건Room Temperature
라벨 또는 염료Bromophenol Blue, Xylene Cyanol
제품 유형Gel Loading Solution
Unit SizeEach
구성 및 보관
Store at -20°C.

자주 묻는 질문(FAQ)

How can generation of replication competent adenoviruses be avoided when using your pSilencer adeno 1.0-CMV System?

Although there is only a very small chance of creating replication competent virus, steps should still be taken to avoid added risk. The virus is expanded in two rounds of amplification, and all secondary expansions should be performed from an initial expansion stock. Secondary amplifications in HEK293 cells should not be performed from the stock of another secondary expansion, as this could increase the chance of making replication competent virus. See the product manual for more detail and guidelines for screening.

Find additional tips, troubleshooting help, and resources within our RNAi Support Center.