Cells-to-Signal™ Lysis Buffer (10 ml Bottle)
Invitrogen™

Cells-to-Signal™ Lysis Buffer (10 ml Bottle)

The same lysis buffer found in the Ambion® Cells-to-Signal™ Kit. Supplied in 5 bottles containing 10 mL each.Read more
Have Questions?
Catalog NumberQuantity
AM87285 x 10 mL bottle
Catalog number AM8728
Price (USD)
1,498.00
Each
Quantity:
5 x 10 mL bottle
Price (USD)
1,498.00
Each
The same lysis buffer found in the Ambion® Cells-to-Signal™ Kit. Supplied in 5 bottles containing 10 mL each.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Product LineAmbion, Cells-to-Signal
Product TypeCell Lysis Buffer
Quantity5 x 10 mL bottle
Shipping ConditionRoom Temperature
Unit SizeEach
Contents & Storage
Lysis Buffer may be stored at any temperature.

Frequently asked questions (FAQs)

When should I use Cells-to-Signal lysis vs. purified RNA for the Clariom GO Screen Assay?

Generally, the more samples you are running for the Clariom GO Screen Assay, the more time and cost will be saved with direct Cells-to-Signal lysis.

Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.

Which cell lines can I lyse with the Cells-to-Signal Kit for use with the Clariom GO Screen Assay?

Two cell lines have been fully verified for the Clariom GO Screen assay, HepG2 and HeLa. General guidance for creating lysates with other cell lines is included in the user guide (https://assets.thermofisher.com/TFS-Assets/LSG/manuals/MAN0018542_ClariomGO_Screen384HT_UG.pdf).

Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.

How do I order the Clariom GO Screen Assay?

You can purchase the Clariom GO Screen Assay (Cat. No. 952361). To start a GO Screen project, contact your local representative and/or complete the quote request form via the product page (https://www.thermofisher.com/order/catalog/product/952361). Our verified service providers are available to help you run your GO Screen assay and can assist with the management of your project.

Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.