Ammonium Acetate (5 M), RNase-free, 100 mL - FAQs
Ammonium Acetate (5 M), RNase-free, 100 mL - FAQs
View additional product information for Ammonium Acetate (5 M), RNase-free - FAQs (AM9070G, AM9071)
1 product FAQs found
What salts can be used to precipitate total RNA? What are the advantages of each? Do they require ethanol?
The three salts that can be used are:
- Guanidine thiocyanate, which requires ethanol. Guanidine thiocyanate is a common agent used for isolating RNA and we recommend it especially for tissues high in ribonuclease activity, such as the pancreas or spleen.
- Ammonium acetate, which requires ethanol. Ammonium acetate is useful when reducing coprecipitation of unwanted dNTPs and oligosaccharides. However, it should not be used when the nucleic acid will be phosphorylated using T4 polynucleotide kinase, since this enzyme is inhibited by ammonium ions.
- Lithium chloride, which does not require ethanol. LiCl is very effective in precipitating RNA but does not efficiently precipitate protein or DNA. It also does not precipitate unincorporated nucleotides.