LiCl Precipitation Solution (7.5 M)
Invitrogen™

LiCl Precipitation Solution (7.5 M)

Precipitation solution offers major advantages over other RNA precipitation methods in that it does not precipitate DNA, protein, or carbohydrates.
Have Questions?
Catalog NumberQuantity
AM9480100 mL
Catalog number AM9480
Price (USD)
139.65
Online exclusive
149.00
Save 9.35
Each
Quantity:
100 mL
Price (USD)
139.65
Online exclusive
149.00
Save 9.35
Each

Precipitation solution offers major advantages over other RNA precipitation methods in that it does not precipitate DNA, protein, or carbohydrates. It is supplied in one bottle of 100 mL at a concentration of 7.5M. It is the method of choice for removing inhibitors of translation or cDNA synthesis from RNA preparations.

Note: LiCl precipitation may not remove all DNA contamination from total RNA isolations. If the RNA is to be used for RT-PCR, it should be DNase I treated. The solution is rigorously tested for contaminating nonspecific endonuclease, exonuclease, and RNase activity.

For Research Use Only. Not for use in diagnostic procedures.

Specifications
Concentration7.5 M
For Use With (Application)RNA Extraction
Product LineAmbion
PurityDNase-Free, RNase-Free
Quantity100 mL
Shipping ConditionRoom Temperature
Unit SizeEach
Contents & Storage
Store at 2°C to 8°C.