How many freeze thaw cycles do you recommend for Invitrogen, Salmon Sperm DNA, sheared (Cat. No. AM9680)?
We do not have specific guidance on the number of freeze thaws for Salmon Sperm DNA, but it can be frozen and thawed multiple times.
Search
View additional product information for Salmon Sperm DNA, sheared (10 mg/mL) - FAQs (AM9680)
3 product FAQs found