Plant RNA Isolation Aid
Invitrogen™
Plant RNA Isolation Aid
Plant RNA Isolation Aid is recommended for use with the RNAqueous Kit or MagMAX Plant RNA Isolation Kit for purificationRead more
Have Questions?
Catalog NumberQuantity
AM969010 mL
Catalog number AM9690
Price (USD)
120.65
Online exclusive
137.00
Save 16.35
Each
Quantity:
10 mL
Price (USD)
120.65
Online exclusive
137.00
Save 16.35
Each
Plant RNA Isolation Aid is recommended for use with the RNAqueous Kit or MagMAX Plant RNA Isolation Kit for purification of total RNA from plant tissues.
• Removes common plant contaminants such as polyphenolics and polysaccharides
• No phenol extraction, alcohol precipitation, or proteinase digestion steps required when used with the recommended kits
Plant RNA Isolation Aid contains polyvinylpyrrolidone (PVP), a high molecular weight polymer that binds to contaminants such as polyphenolics and polysaccharides that are commonly present in plant tissues. Glass fiber filter–based isolation, employed by the RNAqueous Kit and MagMAX Plant RNA Isolation Kit, is especially suited for plant RNA isolation because it does not include any alcohol precipitation steps; many common contaminants found in plant tissue are known to co-precipitate with nucleic acid during alcohol precipitation. When used with the RNAqueous Kit or MagMAX Plant RNA Isolation Kit, the procedure consists of disrupting the plant tissue in a guanidinium-based lysis solution to which the Plant RNA Isolation Aid has been added. A brief centrifugation then removes the contaminants. The supernatant is passed through a glass fiber filter which binds RNA. The filter is then washed and the RNA is eluted in a small volume (50 μL or more) of elution solution.
Accessory products
The Plant RNA Isolation Aid is designed for use with the RNAqueous Kit (Cat. No. AM1912) or MagMAX Plant RNA Isolation Kit (Cat. No. A33784) for RNA purification from plant tissues.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Final Product TypeRNA Supernatant (for use with RNAqueous™ kit)
For Use With (Equipment)RNAqueous™ Kit
High-throughput CompatibilityNot High-throughput Compatible (Manual)
Quantity10 mL
Shipping ConditionRoom Temperature
Isolation TechnologyPVP Reagent
Sample TypePlant Samples
Unit SizeEach
Contents & Storage
Store Plant RNA Isolation Aid at room temperature or at 4°C.
Frequently asked questions (FAQs)
I used the Plant RNA Isolation Aid along with the RNAqueous kit to isolate total RNA from my plant sample. My RNA preparation is too dilute. What do you recommend?
Yields may be improved in some cases by keeping the preparation more concentrated, especially if the tissue has a high water content. Try reducing the amount of lysis solution. Significant errors in weighing small amounts of tissue (<100 mg) can be introduced if the tissue is extremely wet. Tissue samples should be blotted dry just before weighing. Watery tissues may yield more RNA if they are lyophilized before disruption. This increases the ratio of lysis solution to dry-weight tissue.
What kind of Polyvinylpyrrolidone (PVP) do you recommend using with MagMAX Plant DNA and RNA isolation kits since there are different kinds available?
For the MagMAX Plant DNA Isolation Kit, we recommend using PVP M.W. 40,000, Cat. No. J62417.A1 (https://www.thermofisher.com/order/catalog/product/J62417.A1).
For the MagMAX Plant RNA Isolation Kit, we recommend using Plant RNA Isolation Aid, Cat. No. AM9690 (https://www.thermofisher.com/order/catalog/product/AM9690).