RNaseZap™ RNase Decontamination Wipes, 100 Sheets - FAQs
RNaseZap™ RNase Decontamination Wipes, 100 Sheets - FAQs
View additional product information for RNaseZap™ RNase Decontamination Wipes Refill - FAQs (AM9788, AM9786)
1 product FAQs found
What is the formulation and pH of RNaseZAP Rnase Decontamination Solution (Cat. Nos. AM9782, AM9784, AM9780)?
The formulation for RNaseZAP RNase Decontamination Solution (Cat. Nos. AM9782, AM9784, AM9780) is proprietary. The pH is 7.0-7.4.
Find additional tips, troubleshooting help, and resources within our RNA Sample Collection, Protection, and Isolation Support Center.