SDS, 10% Solution, RNase-free, 500 mL - FAQs
SDS, 10% Solution, RNase-free, 500 mL - FAQs
View additional product information for SDS, 10% Solution, RNase-free - FAQs (AM9822, AM9823)
1 product FAQs found
What does SDS Solution (Cat. No. AM9820, AM9822, AM9823) contain?
Ambion molecular biology grade SDS Solution (Cat. No. AM9820, AM9822, AM9823) consists of Sodium Dodecyl Sulfate (SDS) and water. The solution is certified to be RNase-free and ready-to-use. AM9820: 20% SDS Solution, RNase-free, 250 mL
AM9822: 10% SDS Solution, RNase-free, 500 mL
AM9823: 10% SDS Solution, RNase-free, 1 L