Tris (1 M), pH 7.0, RNase-free
Invitrogen™

Tris (1 M), pH 7.0, RNase-free

Invitrogen Molecular biology grade, 1M Tris, pH 7.0 solution is supplied in one bottle. The buffer is certified RNase-free, economical, and ready-to-use.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
AM9850G100 mL
AM9851500 mL
Catalog number AM9850G
Price (USD)
62.65
Online exclusive
66.25
Save 3.60
Each
Quantity:
100 mL
Price (USD)
62.65
Online exclusive
66.25
Save 3.60
Each

Invitrogen Molecular biology grade, 1M Tris, pH 7.0 solution is supplied in one bottle. The buffer is certified RNase-free, economical, and ready-to-use. Due to the ubiquitous presence of RNases, manufacturing products for use with RNA is especially challenging. Invitrogen's nuclease-free reagents and buffers are manufactured in facilities specifically designed to prevent the introduction of nucleases. Highly sensitive RNase assays are performed at several different stages of the manufacturing process to ensure the highest quality. These reagents are rigorously tested for contaminating nonspecific endonuclease, exonuclease, and RNase activity.

For Research Use Only. Not for use in diagnostic procedures.

Specifications
Formulation1,3-Propanediol, 2-Amino-2-(Hydroxymethyl)-,Hydrochloride
Recommended StorageStore at room temperature.
Concentration1 M
GradeMolecular Biology
Physical FormLiquid
Product LineAmbion
Quantity100 mL
pH7
Unit SizeEach