10X TBE (powder)
Invitrogen™

10X TBE (powder)

Ambion™ 10X TBE (Tris-Borate-EDTA Buffer) is thoroughly tested for RNase and DNase contamination, making it ideal for use in sensitiveRead more
Have Questions?
Catalog NumberQuantity
AM986410 x 1 L
Catalog number AM9864
Price (USD)
466.00
Each
Quantity:
10 x 1 L
Price (USD)
466.00
Each
Ambion™ 10X TBE (Tris-Borate-EDTA Buffer) is thoroughly tested for RNase and DNase contamination, making it ideal for use in sensitive application. 10 packets are provided, each containing the necessary powder to make 1 liter of 10X TBE buffer upon the addition of H2O. These reagents are rigorously tested for contaminating nonspecific endonuclease, exonuclease, and RNase activity.

Specifications:
Appearance: White crystalline powder Molecular Weight: Tris buffer component - 121.14; Boric acid component - 61.83; EDTA component - 372.3
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Chemical Name or MaterialRunning Buffer
Recommended StorageStore at room temperature.
Concentration10X
For Use With (Application)Nucleic acid gel electrophoresis & blotting
Physical FormPowder
Product LineAmbion
Quantity10 x 1 L
Unit SizeEach

Frequently asked questions (FAQs)

Can I autoclave agarose to prepare agarose gels?

No. Autoclaving at temperatures that are too high or pressures that are too high can hydrolyze the agarose polymer and weaken it.

Do you have any tips for preparing high-percentage (3% to 4%) agarose gels?

Add the powder to cold buffer while stirring. Let the agarose rehydrate for 1 to 2 hr and then heat slowly until agarose is completely dissolved. Using a microwave at low power settings is acceptable.

What can I do to prevent static on agarose bottles?

One recommendation: get a supply of fabric softener sheets and wipe the bottle with a sheet before opening it.

How can I avoid the build-up of the precipitous film sometimes seen in 10X TBE?

The precipitation of concentrated TBE stocks may be due to nucleation of salt crystals by dust particles or other insoluble materials. Therefore, filtering the solution through a 0.2 µm cellulose acetate or cellulose nitrate filter after preparation helps avoid this precipitate. [Mayeda A, Krainer A (1991) BioTechniques 10.2, 1820]

How can generation of replication competent adenoviruses be avoided when using your pSilencer adeno 1.0-CMV System?

Although there is only a very small chance of creating replication competent virus, steps should still be taken to avoid added risk. The virus is expanded in two rounds of amplification, and all secondary expansions should be performed from an initial expansion stock. Secondary amplifications in HEK293 cells should not be performed from the stock of another secondary expansion, as this could increase the chance of making replication competent virus. See the product manual for more detail and guidelines for screening.

Find additional tips, troubleshooting help, and resources within our RNAi Support Center.