What are the compositions DNAZap PCR DNA Degradation Solutions (Cat. No. AM9890)?
The compositions of these buffers are proprietary.
Find additional tips, troubleshooting help, and resources within our PCR and cDNA Synthesis Support Center.
Find additional tips, troubleshooting help, and resources within our PCR and cDNA Synthesis Support Center.