Nuclease-Free Water (not DEPC-Treated), 10 x 50 mL - FAQs
Nuclease-Free Water (not DEPC-Treated), 10 x 50 mL - FAQs
View additional product information for Nuclease-Free Water (not DEPC-Treated) - FAQs (AM9939, AM9937, AM9938, 4387936, AM9932, AM9930)
2 product FAQs found
What is the expiration date for Nuclease-Free Water (Cat. No. AM9937)?
This will be covered by our standard 12-month warranty as outlined in Section 8.1 of our Terms and Conditions of Sale. The 12-month warranty period begins from the date we ship the product to you. If your warranty has expired, the product may still perform as expected (when stored properly), we are just not able to guarantee or issue free-of-charge replacements in the case of poor performance.
Find additional tips, troubleshooting help, and resources within our Nucleic Acid Purification and Analysis Support Center.
I would like to order a TaqMan SNP Genotyping Assay. Does the kit include everything I need to perform my experiment or do I need to purchase reagents separately?
Each predesigned TaqMan SNP Genotyping Assay is delivered in a single tube consisting of two differentially labeled allele-specific TaqMan MGB (minor groove binding) probes and a pair of PCR primers. For more information, please see the following link: https://www.thermofisher.com/us/en/home/life-science/pcr/real-time-pcr/real-time-pcr-applications/genetic-variation-analysis-using-real-time/snp-genotyping-with-real-time-pcr.html
Besides the TaqMan SNP Genotyping Assay, you would need to purchase a Genotyping Master Mix and nuclease-free water. A list of compatible master mixes can be found in the following link: https://assets.thermofisher.com/TFS-Assets/LSG/manuals/MAN0009593_TaqManSNP_UG.pdf.
Find additional tips, troubleshooting help, and resources within our Gene Expression Analysis & Genotyping Support Center.