MEGAscript™ T7 Transcription Kit, 200 reactions - FAQs
MEGAscript™ T7 Transcription Kit, 200 reactions - FAQs
View additional product information for MEGAscript™ T7 Transcription Kit - FAQs (AM1334, AM1333, AMB13345)
1 product FAQs found
With the MEGAscript T7 Transcription Kit, can the T7 polymerase synthesize RNA from a single-stranded DNA template, containing the T7 promoter sequence, or does it require a double-stranded DNA promoter sequence to work?
The T7 polymerase requires, at the minimum, for the promoter sequence to be double-stranded. The rest of the template does not necessarily have to be double-stranded.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.