EVOS™ Light Cube, AO - FAQs
EVOS™ Light Cube, AO - FAQs
View additional product information for EVOS™ Light Cube, AO - FAQs (AMEP4670)
1 product FAQs found
What is the difference between the GFP light cube and the Qdot 525 light cube as they both allow detection of green fluorescence?
The GFP light cube (Ex = 470/22; Em = 510/42) has excitation centered at 470 nm covering 11 nm on each side for a total excitation range from 459 to 481 nm. Its emission is centered at 510 nm with 21 nm on each side for a total emission bandpass range from 489 to 531 nm.
The Qdot 525 light cube (Ex = 445/45; Em = 525/15) has excitation at 445 nm with 22.5 nm on each side for a total excitation range from 422.5 to 467.5. Its emission is centered at 525 nm with 7.5 nm on each side for a total emission bandpass range from 517.5 to 532.5.
The GFP light cube is best suited for fluorophores that are efficiently excited from 460 to 480 nm and emission with in the blue-green to green range.
The Qdot 525 light cube is better suited for exciting quantum dots at lower wavelengths (higher energy) and provides a narrower emission in the green range.