EVOS™ Light Cube, TagBFP 2.0 - FAQs
EVOS™ Light Cube, TagBFP 2.0 - FAQs
View additional product information for EVOS™ Light Cube, TagBFP 2.0 - FAQs (AMEP4968)
2 product FAQs found
Are EVOS 2.0 Light Cubes compatible with the Countess II FL?
Yes. All our EVOS Light Cubes can be used with the Countess II FL.
Are EVOS 2.0 Light Cubes compatible with older EVOS instruments?
Yes. Our EVOS 2.0 Light cubes can be used in all EVOS instruments with EVOS light cube functionality.