EVOS™ 4X Objective, fluorite, LWD, phase-contrast, 0.13NA/10.58WD - FAQs
EVOS™ 4X Objective, fluorite, LWD, phase-contrast, 0.13NA/10.58WD - FAQs
View additional product information for EVOS™ 4X Objective, fluorite, LWD, phase-contrast, 0.13NA/10.58WD - FAQs (AMEP4980)
1 product FAQs found
Do you offer alternatives for the discontinued objective lenses, EVOS 20X Objective (Cat. No. AMEP4682) and EVOS 10X Objective (Cat. No. AMEP4682), that will fit the EVOS FL Imaging System?
We offer improved versions of those lenses listed below:
- EVOS 20X Objective, fluorite, LWD, phase contrast, 0.45 NA/6.12WD (Cat. No. AMEP4982)
- EVOS 10X Objective, fluorite, LWD, phase-contrast, 0.30NA/7.13WD (Cat. No. AMEP4981)
The 4X objective with similar specs as the ones above is:
- EVOS 4X Objective, fluorite, LWD, phase-contrast, 0.13NA/10.58WD (Cat. No. AMEP4980)
However, depending on the applications you have in mind for the 4X objective, you can also consider:
- EVOS 4X Objective, fluorite, LWD, 0.13NA/10.58WD (Cat. No. AMEP4922)
- EVOS 4X Objective, achromat, LWD, phase-contrast, 0.13NA/10.58WD (Cat. No. AMEP4932)