Countess™ 3 Automated Cell Counter, 1 instrument - FAQs
Countess™ 3 Automated Cell Counter, 1 instrument - FAQs
View additional product information for Countess™ 3 Automated Cell Counter - FAQs (A49865, A50302, AMQAX2000, A49891, A50298, A49862)
7 product FAQs found
Why am I getting a much higher bead count on the Countess 3 Standard Slide (Cat. No. A51876)?
When counting cells that have been mixed with Trypan Blue, the Countess accounts for this 1:1 dilution and multiplies the count by 2.
When using the Countess 3 Standard Slide, the Trypan Blue correction box needs to be unchecked, since a 1:1 dilution was not made with Trypan Blue.
What cell counter do you offer that can count live and dead cells, measure cell viability, cell size, has high-resolution touch screen, and comes with reusable slides?
Are there any recommended parameter settings on the Countess 3/3FL Automated Cell Counter (Cat. Nos. AMQAF2000, AMQAX2000) for specific cell lines?
No. The Countess 3/3 FL Automated Cell Counter (Cat. Nos. AMQAF2000, AMQAX2000) parameter settings, especially brightness, can vary between instruments due to slight differences in lamp output and other factors.
What can we do with the FCS file generated by the Countess 3/3 FL Automated Cell Counter (Cat. Nos. AMQAF2000, AMQAX2000)?
The Countess 3/3FL Automated Cell Counter (Cat. Nos. AMQAF2000, AMQAX2000) FCS files can be uploaded to most flow cytometry software that can accept FCS files, such as FlowJo or the Attune NxT software. Using the flow cytometry software, you may see the distribution of all cells and adjust/fine-tune parameter settings for only the cells of interest or analyze outliers.
Can we use any EVOS Light Cube with the Countess 3 FL Automated Cell Counter (Cat. No. AMQAF2000)?
With the exception of EVOS Light Cube, White (Cat no. AMEP4672) any of the EVOS Light Cubes for fluorescence detection can be used on the Countess 3 FL Automated Cell Counter (Cat. No. AMQAF2000).
Can I use crystal violet with the Countess 3 (Cat. No. AMQAX2000) and 3 FL Automated Cell Counter (Cat. No. AMQAF2000)?
Yes. Crystal violet is compatible with Countess 3 (Cat. No. AMQAX2000) and 3 FL Automated Cell Counter (AMQAF2000) under brightfield mode detection.
Find additional tips, troubleshooting help, and resources within our Cell Viability, Proliferation, Cryopreservation, and Apoptosis Support Center.
Can I count nuclei with the Countess 3 Automated Cell Counter (Cat. No. AMQAX2000) or Countess 3 FL Automated Cell Counter (Cat. No. AMQAF2000)?
Yes, with software update version 1.3.1324.552, nuclei can be counted using the Countess 3 FL Automated Cell Counter. We recommend performing FL-based counting. Please refer to this application note for detailed instructions.
Find additional tips, troubleshooting help, and resources within our Cell Counting Support Center.