AP2/AP5 Series Alpha Scintillation Probes
El producto real puede variar
AP2/AP5 Series Alpha Scintillation Probes
AP2/AP5 Series Alpha Scintillation Probes
Thermo Scientific™
AP2/AP5 Series Alpha Scintillation Probes
Reliably locate alpha contamination on people or on workplace surfaces with ruggedly built, easy to handle zinc sulfide scintillator instruments, the preferred technology.
Have Questions?
Número de catálogoDescripción
AP5RBSondas de centelleo alfa serie AP5/RB
Número de catálogo AP5RB
Precio (MXN)
-
Descripción:
Sondas de centelleo alfa serie AP5/RB
Conclusively locate alpha contamination on personnel or workplace surfaces. Thermo Scientific™ AP2 and AP5 Series Alpha Scintillation Probes are general purpose instruments containing a zinc sulfide alpha scintillator, the preferred technology for detecting alpha contamination on the hands, feet or other surfaces. The AP2 probe has a conveniently angled radiation window and is contained in a light alloy housing for easy self monitoring. The AP5 probe features rugged construction and field-replaceable radiation windows for reliable operation while monitoring workplaces, benches, personnel and tools.
AP2 Series
50cm2 square window.
The low weight and durability of the probes make them easy to handle.
Conveniently angled radiation window for self monitoring of clothing.
Field-replaceable radiation windows.
Available versions:
AP2/4A with connector type PET
AP2/4AR with connector type PET, ruggedized version
AP2/4B with connector type MHV
AP5 Series
100cm2 rectangular window.
High transmission rectangular hex-grille.
Slim, lightweight, wedge-shaped profile.
Field-replaceable radiation windows.
Available versions:
AP5/AD with connector type PET.
AP5/RA with connector type PET, ruggedized version.
AP5/BD with connector type MHV.
AP5/RB with connector type MHV, ruggedized version.
Information on calibration sources can be provided on request.
Especificaciones
DescripciónSondas de centelleo alfa serie AP5/RB
Peso (imperial)1.1 lb.
Tamaño de la ventana100 cm2
Medición de fondos-1: ≤0.1
Eficacia23% 241 Am (AP5RB)
Tipo de productoAlpha Scintillation Probe
Peso (métrico)0.5 kg
Unit SizeEach