This product is now part of a food safety range that includes Thermo Scientific™ culture media, molecular solutions, and other products for your complete microbiological workflow. In US, Canada and Europe, order this product from Thermo Fisher Scientific microbiology. >> Order now
Applied Biosystems™

Pathatrix™ Dual (Listeria/Salmonella spp.) Kit

Catalog number:  APD50

Back to top

Description

The Pathatrix® DUAL (Listeria/Salmonella spp.) Kit, when used with the Pathatrix® Auto Instrument, facilitates the concentration and isolation of Listeria spp. and Salmonella spp. bacteria from pre-enriched food and environmental samples for downstream detection by PCR, immunoassay or culture.

When used with the Pathatrix® Auto Instrument, the Pathatrix® DUAL (Listeria/Salmonella spp.) Kit:
• Reduces time-to-results – concentrates target organisms, so less enrichment time is required
• Minimizes false positive and indeterminate rates– isolates specific bacteria from background flora and PCR inhibitors

Each kit includes one vial of antibody-coated beads and sufficient numbers of sterile, single-use plastic consumables for 50 samples.
For Testing of Food and Environmental Samples Only.

Specifications

Final Product: Bacteria (Intact)
For Use With (Equipment): Pathatrix® Auto System
High Throughput Compatibility: Automated Protocols,High Throughput-Compatible
Isolation Technology: Immunomagnetic Separation
Number of Reactions: 50 Tests
Product Line: Pathatrix™
Product Size: 50 tests
Sample Source: Individual Samples
Sample Treatment: Overnight Incubation (Pre-Enrichment)
Sample Type (Specific): Environmental Sample,Food
Sample Volume: 50 mL
Strain or Genotype: Listeria spp,Salmonella spp.
Target Organism Class: Listeria spp.,Salmonella spp.

Contents & storage

Each kit includes one vial of antibody-coated beads and sufficient sterile, single-use plastic consumables for 50 samples.

Store beads at 2–8°C; do not freeze.