This product is now part of a food safety range that includes Thermo Scientific™ culture media, molecular solutions, and other products for your complete microbiological workflow. In US, Canada and Europe, order this product from Thermo Fisher Scientific microbiology. >> Order now
Applied Biosystems™

Pathatrix™ Cronobacter spp. Kit, same day

Catalog number:  APSAK50

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The Pathatrix® Cronobacter spp. Kit – Same Day, when used with the Pathatrix® Auto Instrument, facilitates the concentration of Cronobacter bacteria from pre-enriched food and environmental samples for downstream detection by PCR, immunoassay or culture. The kit is optimized for use with protocols that require 8 or fewer hours of pre-enrichment time.

Each kit includes one vial of antibody-coated beads and sufficient numbers of sterile, single-use plastic consumables for 50 samples.
For Testing of Food and Environmental Samples Only.

Specifications

Final Product: Bacteria (Intact)
For Use With (Equipment): Pathatrix® Auto System
High Throughput Compatibility: Automated Protocols, High Throughput-Compatible
Isolation Technology: Immunomagnetic Separation
Number of Reactions: 50 Tests
Product Line: Pathatrix™
Product Size: 50 tests
Sample Source: Individual Samples
Sample Treatment: Overnight Incubation (Pre-Enrichment), Same-Day Incubation (Pre-Enrichment)
Sample Type (Specific): Environmental Sample, Food
Sample Volume: 50 mL
Strain or Genotype: Cronobacter spp.
Target Organism Class: Cronobacter spp.

Contents & storage

Each kit includes one vial of antibody-coated beads and sufficient sterile, single-use plastic consumables for 50 samples.

Store beads at 2–8°C; do not freeze.

Documents

Manuals & protocols