Sensititre™ Veterinary Aquatic Animals AQUA2 Plate
Thermo Scientific™

Sensititre™ Veterinary Aquatic Animals AQUA2 Plate

Sensititre™ AQUA2 plate delivers accurate MIC results with gold-standard broth microdilution for aquatic animal susceptibility testing.
Have Questions?
Catalog NumberAntimicrobial Agents
AQUA2Various
Catalog number AQUA2
Price (USD)
-
Antimicrobial Agents:
Various
Antimicrobial susceptibility testing plate delivering host-animal-specific results in a high-quality, ready to use AST broth microdilution (BMD) plate format, meeting the latest CLSI guidelines.

The Thermo Scientific™ Sensititre™ AQUA2 plate delivers host-animal-specific results by utilizing broth microdilution (BMD), the gold standard1 phenotypic method for Antimicrobial Susceptibility Testing (AST). Get access to one of the most comprehensive range of antimicrobials on a single plate format, designed for veterinary use only. 

  • Veterinary Use Only (VET)
  • BMD provides accurate MIC results therefore negating the need for repeat testing
  • Designed for use with CLSI guidelines
  • Sensititre™ MIC determination is achieved using direct measurement rather than relying on extrapolated data, thereby providing an accurate MIC result, not an estimate
  • All contained antibiotics are also available in customizable plate options, along with access to over 300 antimicrobials including veterinary specific antimicrobials, to enable meeting local formulary and stewardship program requirements 

Thermo Fisher Scientific is a leader in providing AST solutions that help in the fight against antimicrobial resistance, offering high performance automated and manual testing options and one of the largest, most up-to-date selections of antimicrobials for susceptibility testing.

The Sensititre™ System utilizes gold standard-level1 MIC accuracy, providing superior quality and reproducibility for accurate results the first time.

For Veterinary Use Only.

¹Jayol A, Nordmann P, André C, Poirel L, Dubois V. Evaluation of three broth microdilution systems to determine colistin susceptibility of Gram-negative bacilli. J Antimicrob Chemother. 2018; 73: 1272-1278. PubMed

Specifications
Antimicrobial AgentsVarious
For Use With (Application)Sensititre™
Format190 x 200 x 200 mm (H x W x D) plate
No. per Pack10
Packaging TypeBox
Shelf Life2 years
Product TypeAST Plate
SpeciesAquatic Animals
Target Organism ClassAquatic Bacterial Isolates
Test TypeVeterinary AST, Minimum Inhibitory Concentration (MIC)
Unit SizeEach