See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
New Products
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
Home
▸
Shop All Products
▸
Ovens and Furnaces
▸
Furnaces
▸
Muffle Furnaces
▸
Thermolyne™ Atmosphere Controlled Ashing Furnace Accessories
Thermo Scientific™
Thermolyne™ Atmosphere Controlled Ashing Furnace Accessories
Use these accessories with Thermolyne Atmosphere Controlled Ashing Furnaces.
Have Questions?
Change view
Catalog Number
Type
For Use With (Equipment)
AY408X1
Inconel Manifold
F6000 Furnace
AY408X1A
Exhaust Tubing Kit
Ashing and Muffle Furnaces
AY718X1
Inconel Manifold
F30400 Furnace
3 Options
Catalog number
AY408X1
Price (USD)
-
Request A Quote
Type:
Inconel Manifold
For Use With (Equipment):
F6000 Furnace
Use Thermo Scientific™ Thermolyne™ Atmosphere Controlled Ashing Furnace Accessories to further the flexibility of your ashing furnace.
Specifications
Description
Manifold; For F6000 furnace
Style
Inconel Manifold
For Use With (Equipment)
F6000 Furnace
Temperature (Metric)
1093°C
Type
Inconel Manifold