Transfer Vessels Withdrawal Device
Thermo Scientific™
Transfer Vessels Withdrawal Device
Easily dispense up to 8L/min. (2.12 gal./min.) without pouring using the Thermo Scientific™ Transfer Vessels Withdrawal Device.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFor Use With (Equipment)
AY509X3LN2 Transfer Vessels, For Thermo 30
AY509X4LN2 Transfer Vessels, For Thermo 20
AY509X5LN2 Transfer Vessels, For Thermo 10
Catalog number AY509X3
Price (USD)
-
For Use With (Equipment):
LN2 Transfer Vessels, For Thermo 30
Easily dispense up to 8L/min. (2.12 gal./min.) without pouring using the Thermo Scientific™ Transfer Vessels Withdrawal Device.
  • Available for 10, 20 and 30L models
  • Warranty varies by country, please contact us for details
  • Specifications
    For Use With (Equipment)LN2 Transfer Vessels, For Thermo 30
    TypeLN2 Withdrawal Device, 8L/min.
    Capacity (English)1.05 cu. ft.
    Capacity (Metric)30 L
    Unit SizeEach