Please note: order placement quantity must be in multiples of 20, minimum of 20 required.
Thermo Scientific™ 2D Tank Liner, ASI™ 26 film
Thermo Scientific™

Thermo Scientific™ 2D Tank Liner, ASI™ 26 film

The 560 L Thermo Scientific™ 2D Tank Liner System without bottom drain is composed of Thermo Scientific™ ASI™ 26 PolyethyleneRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Tubing Style
B100050I
also known as B100050-I
400 LNo Tubing
B100051I
also known as B100051-I
560 LNo Tubing
B100048I
also known as B100048-I
50 LNo Tubing
B100049I340 LNo Tubing
B100038I
also known as B100038-I
130 LNo Tubing
B100037I
also known as B100037-I
200 LNo Tubing
Catalog number B100050I
also known as B100050-I
Price (USD)
-
Capacity (Metric):
400 L
Tubing Style:
No Tubing
The 560 L Thermo Scientific™ 2D Tank Liner System without bottom drain is composed of Thermo Scientific™ ASI™ 26 Polyethylene Film, a durable 5.0 mL film, with dimensions optimized for Thermo Scientific™ drums and commonly-used, industry standard cylindrical tanks. Standard Thermo Scientific tank liners feature an open top for maximum recovery, with drum dolly available to provide mobility.

Features
• Minimizes bioburden due to construction under sterile cGMP conditions
• Eliminates post-use cleaning steps required with reusable containers, helping to reduce cross-contamination risks

Applications
• Removes the need for tank cleaning and helps to reduce cycle times
• Facilitates hydration of powdered media and buffers, and pooling of non-sterile solutions and fluids

Specifications
Dimensions71.1 cm x 111.8 cm (28 in. x 44 in.)
For Use With (Equipment)SV30115
Product LineHyPerforma
Quantity400 L, 20 per case
Shipping ConditionRoom Temperature
Sterilization MethodIrradiated
Capacity (Metric)400 L
Film DescriptionASI 26
MaterialPolyethylene
Port Detail0 Ports
Tubing StyleNo Tubing
Unit SizeEach
Contents & Storage
Store at room temperature.