Please note: order placement quantity must be in multiples of 40, minimum of 40 required.
Thermo Scientific™ 3D Tank Liner, ASI™ 26 film
Thermo Scientific™

Thermo Scientific™ 3D Tank Liner, ASI™ 26 film

The 560 L Thermo Scientific™ 3D Tank Liner System without bottom drain is composed of Thermo Scientific™ ASI™ 26 PolyethyleneRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Dimensions
B100364I
also known as B100364-I
50 L33 cm x 68.6 cm (13 in. x 27 in.)
B100363I
also known as B100363-I
200 L55.9 cm x 91.4 cm (22 in. x 36 in.)
B100373I
also known as B100373-I
100 L45.7 cm x 76.2 cm (18 in. x 30 in.)
B101459I
also known as B101459-I
300 L70 cm x 122 cm (24 in. x 48 in.)
B101460I
also known as B101460-I
560 L78.7 cm x 124.5 cm (31 in. x 49 in.)
Catalog number B100364I
also known as B100364-I
Price (USD)
-
Capacity (Metric):
50 L
Dimensions:
33 cm x 68.6 cm (13 in. x 27 in.)
The 560 L Thermo Scientific™ 3D Tank Liner System without bottom drain is composed of Thermo Scientific™ ASI™ 26 Polyethylene Film, a durable 5.0 mL film, with dimensions optimized for Thermo Scientific™ drums and commonly-used, industry standard cylindrical tanks. Standard Thermo Scientific tank liners feature an open top for maximum recovery, with drum dolly available to provide mobility.

Features
• Minimizes bioburden due to construction under sterile cGMP conditions
• Eliminates post-use cleaning steps required with reusable containers, helping to reduce cross-contamination risks

Applications
• Removes the need for tank cleaning and helps to reduce cycle times
• Facilitates hydration of powdered media and buffers, and pooling of non-sterile solutions and fluids

Specifications
Dimensions33 cm x 68.6 cm (13 in. x 27 in.)
Outer PackagingIndividually packed in a header bag
Product LineHyPerforma
Quantity40 per case
Shipping ConditionRoom Temperature
Sterilization MethodIrradiated
Capacity (Metric)50 L
Film DescriptionASI 26
MaterialPolyethylene
Port Detail0 Ports
Tubing StyleNo Tubing
Unit Size50 L
Contents & Storage
Store at room temperature.