Alpha-Bungarotoxin Conjugates
Invitrogen™

Alpha-Bungarotoxin Conjugates

Thermo Fisher Scientific offers a broad selection of Invitrogen alpha-bungarotoxin conjugates, including Alexa Fluor and Alexa Fluor Plus conjugates.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLabel or Dye
B1601Unlabeled
B56130Alexa Fluor™ Plus 405
B13422Alexa Fluor™ 488
B35451Alexa Fluor™ 555
B13423Alexa Fluor™ 594
B35450Alexa Fluor™ 647
B1196Biotin-XX
T1175Tetramethylrhodamine
Catalog number B1601
Price (USD)
142.65
Online exclusive
152.00
Save 9.35
Each
Label or Dye:
Unlabeled
Price (USD)
142.65
Online exclusive
152.00
Save 9.35
Each
Thermo Fisher Scientific offers a broad selection of Invitrogen alpha-bungarotoxin conjugates, including Alexa Fluor and Alexa Fluor Plus conjugates. Alpha-bungarotoxin, a 74-amino acid peptide extracted from Bungarus multicinctus venom, binds with high affinity to the alpha subunit of the nicotinic acetylcholine receptor (AChR) of neuromuscular junctions.

Our bright and photostable Alexa Fluor 555 alpha-bungarotoxin is a superior choice of probe for visualizing this receptor. Fluorescent alpha-bungarotoxin conjugates can be used to facilitate identification of nicotinic AChRs and localization of neuromuscular junctions. Conjugated alpha-bungarotoxin has been used in variety of applications, including immunofluorescence (IF), immunohistochemistry (IHC), and flow cytometry (FC).

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Label or DyeUnlabeled
Product TypeConjugate
Protein SubtypeOther Proteins
Quantity1 mg
Shipping ConditionRoom Temperature
Product LineShuttle PIP
Unit SizeEach
Contents & Storage
Store in freezer (–5° to –30°C).

Citations & References (86)

Citations & References
Abstract
Precise localization of alpha7 nicotinic acetylcholine receptors on glutamatergic axon terminals in the rat ventral tegmental area.
Authors:Jones IW, Wonnacott S
Journal:J Neurosci
PubMed ID:15601930
'Alpha7 neuronal nicotinic acetylcholine receptors (nAChRs) constitute one of the predominant nAChR subtypes in the mammalian brain. Within the ventral tegmental area (VTA), nicotine application, paired with postsynaptic stimulation, contributes to a form of long-term potentiation, an effect attributed to presynaptic alpha7 nAChRs on glutamatergic afferents (Mansvelder and McGehee, 2000). ... More
Imaging of receptor trafficking by using alpha-bungarotoxin-binding-site-tagged receptors.
Authors:Sekine-Aizawa Y, Huganir RL,
Journal:Proc Natl Acad Sci U S A
PubMed ID:15563595
'alpha-Amino-3-hydroxy-5-methyl-4-isoxazole-propionate (AMPA) receptors mediate excitatory synaptic transmission and are dynamically regulated during synaptic plasticity in the CNS. The membrane trafficking of AMPA receptors to synapses is critical for the regulation of the efficacy of excitatory synaptic transmission. Direct imaging of AMPA receptors in various cell compartments is important to dissecting ... More
alpha-Bungarotoxin binding sites in rat hippocampal and cortical cultures: initial characterisation, colocalisation with alpha 7 subunits and up-regulation by chronic nicotine treatment.
Authors:Barrantes GE, Rogers AT, Lindstrom J, Wonnacott S
Journal:Brain Res
PubMed ID:7749744
'High density neuronal cultures from rat E18 hippocampus and cortex have been characterised with respect to cholinergic binding sites. No specific binding of [3H]nicotine or [3H]cytisine to live cells in situ was detected although the limit for detection was estimated to be 30 fmol/mg protein. Muscarinic binding sites labelled with ... More
The short-neurotoxin-binding regions on the alpha-chain of human and Torpedo californica acetylcholine receptors.
Authors:Ruan KH, Stiles BG, Atassi MZ
Journal:Biochem J
PubMed ID:2012611
'The continuous regions for short-neurotoxin binding on the alpha-chains of Torpedo californica (electric ray) and human acetylcholine receptors (AChR) were localized by reaction of 125I-labelled cobrotoxin (Cot) and erabutoxin b (Eb) with synthetic overlapping peptides spanning the entire extracellular part of the respective alpha-chains. On Torpedo AChR, five Cot-binding regions ... More
Actions of vesamicol on an alpha-bungarotoxin-sensitive neuronal nicotinic acetylcholine receptor.
Authors:Buckingham SD, Lummis SC, Balk ML, Schroeder M, Sattelle DB
Journal:J Exp Biol
PubMed ID:8228781
'Electrophysiology and binding studies were used to determine the actions of vesamicol [2-(4-phenylpiperidino)cyclohexanol (AH5183)] on an alpha-bungarotoxin-sensitive, neuronal nicotinic acetylcholine receptor in the nervous system of the cockroach, Periplaneta americana. Electrophysiological studies on an identified motor neurone revealed a reversible blocking action of (+/-)-vesamicol on the response to ionophoretically applied ... More