Owl™ EasyCast™ B1A Mini Gel and A2-OK Multi Gel Electrophoresis System Combs
Actual product may vary
Owl™ EasyCast™ B1A Mini Gel and A2-OK Multi Gel Electrophoresis System Combs
Owl™ EasyCast™ B1A Mini Gel and A2-OK Multi Gel Electrophoresis System Combs
Thermo Scientific™
Owl™ EasyCast™ B1A Mini Gel and A2-OK Multi Gel Electrophoresis System Combs
Eliminate comb assembly with this assortment of heavy-duty, one-piece combs.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTypeTeeth CountThickness (Metric) Teeth
B1A-5DComb51.5 mm
B1A-5CComb51 mm
B1A-6Double-sided Comb61.0/1.5 mm
B1A-8Double-sided Comb81.0/1.5 mm
B1A-PREPPrep Comb21.5 mm
Catalog number B1A-5D
Price (USD)
80.28
Online exclusive
89.20
Save 8.92
Each
Type:
Comb
Teeth Count:
5
Thickness (Metric) Teeth:
1.5 mm
Price (USD)
80.28
Online exclusive
89.20
Save 8.92
Each
Eliminate comb assembly with Thermo Scientific™ Owl™ EasyCast™ B1A Mini Gel and A2-OK Multi Gel Electrophoresis System Combs. This assortment of heavy-duty, one-piece combs is for use with the EasyCast B1A Mini Gel and A2-OK Multi Gel Electrophoresis System.
Ordering Information:
Customized combs are available (order SKU#: XCM). Please contact your customer service or sales representative for further details.
Specifications
Teeth Count5
Thickness (Metric) Teeth1.5 mm
For Use With (Equipment)Owl™ EasyCast B1A Mini Gel and A2-OK Multi Gel Electrophoresis System
Gel SizeMini
Product LineOwl, EasyCast
TypeComb
Unit SizeEach