4-Sulfonamidophenylhydrazine hydrochloride, 97%
4-Sulfonamidophenylhydrazine hydrochloride, 97%
4-Sulfonamidophenylhydrazine hydrochloride, 97%
Thermo Scientific Chemicals

4-Sulfonamidophenylhydrazine hydrochloride, 97%

CAS: 17852-52-7 | C6H10ClN3O2S | 223.675 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 g
10 g
50 g
Catalog number B20015.09
also known as B20015-09
Price (USD)
43.65
Special offer
Online exclusive
Ends: 30-Jun-2026
51.20
Save 7.55 (15%)
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
43.65
Special offer
Online exclusive
Ends: 30-Jun-2026
51.20
Save 7.55 (15%)
Each
Chemical Identifiers
CAS17852-52-7
IUPAC Namehydrogen 4-hydrazinylbenzene-1-sulfonamide chloride
Molecular FormulaC6H10ClN3O2S
InChI KeyIKEURONJLPUALY-UHFFFAOYSA-N
SMILES[H+].[Cl-].NNC1=CC=C(C=C1)S(N)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to pale brown or pale yellow
FormCrystals or powder or crystalline powder or flakes
Assay (Titration ex Chloride)≥96.0 to ≤104.0%
It is a potentially useful intermediate in the production of Celebrex an anti inflammatory drug.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a potentially useful intermediate in the production of Celebrex an anti inflammatory drug.

Solubility
Soluble in water.

Notes
Hygroscopic. Store away from oxidizing agents, water/moisture and bases. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only
Frank Schönberger, et al.Simple and sensitive method for the determination of celecoxib in human serum by high-performance liquid chromatography with fluorescence detection.Journal of Chromatography B.2002,768(2), 255-260.