Azoxybenzene, 98+%, Thermo Scientific Chemicals
Azoxybenzene, 98+%, Thermo Scientific Chemicals
Azoxybenzene, 98+%, Thermo Scientific Chemicals
Azoxybenzene, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Azoxybenzene, 98+%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
B20021.0910 g
Catalog number B20021.09
Price (USD)
46.20
Each
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)
46.20
Each
Add to cart
Chemical Identifiers
CAS495-48-7
IUPAC Name1-oxo-1,2-diphenylhydrazin-1-ium-2-ide
Molecular FormulaC12H10N2O
InChI KeyGAUZCKBSTZFWCT-UHFFFAOYSA-N
SMILESO=[N+]([N-]C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥98.0%
Melting Point (clear melt)32.0-39.0?C
Appearance (Color)Yellow to orange
FormCrystals or powder or crystalline powder or fused solid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only