2,4-Dichloroanisole, 99%
2,4-Dichloroanisole, 99%
2,4-Dichloroanisole, 99%
Thermo Scientific Chemicals

2,4-Dichloroanisole, 99%

CAS: 553-82-2 | C7H6Cl2O | 177.02 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number B20042.06
also known as B20042-06
Price (USD)
25.00
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
25.00
Each
Chemical Identifiers
CAS553-82-2
IUPAC Name2,4-dichloro-1-methoxybenzene
Molecular FormulaC7H6Cl2O
InChI KeyCICQUFBZCADHHX-UHFFFAOYSA-N
SMILESCOC1=CC=C(Cl)C=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Refractive Index1.5590-1.5630 @ 20°C
Assay (GC)≥98.5%
Appearance (Color)Clear colorless to pale brown
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only