5-Amino-3-tert-butyl-1-methylpyrazole, 98%
5-Amino-3-tert-butyl-1-methylpyrazole, 98%
5-Amino-3-tert-butyl-1-methylpyrazole, 98%
Thermo Scientific Chemicals

5-Amino-3-tert-butyl-1-methylpyrazole, 98%

CAS: 118430-73-2 | C8H15N3 | 153.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number B20095.06
also known as B20095-06
Price (USD)
228.65
Special offer
Online exclusive
Ends: 30-Jun-2026
269.00
Save 40.35 (15%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
228.65
Special offer
Online exclusive
Ends: 30-Jun-2026
269.00
Save 40.35 (15%)
Each
Chemical Identifiers
CAS118430-73-2
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Melting Point (clear melt)153.0-159.0?C
Appearance (Color)White to pale cream or pink
Assay (GC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only