2,3-Diphenylquinoxaline, 98+%
2,3-Diphenylquinoxaline, 98+%
2,3-Diphenylquinoxaline, 98+%
Thermo Scientific Chemicals

2,3-Diphenylquinoxaline, 98+%

CAS: 1684-14-6 | C20H14N2 | 282.346 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number B20100.06
also known as B20100-06
Price (USD)
27.65
Online Exclusive
31.00
Save 3.35 (11%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
27.65
Online Exclusive
31.00
Save 3.35 (11%)
Each
Chemical Identifiers
CAS1684-14-6
IUPAC Name2,3-diphenylquinoxaline
Molecular FormulaC20H14N2
InChI KeyRSNQVABHABAKEZ-UHFFFAOYSA-N
SMILESC1=CC=C(C=C1)C1=C(N=C2C=CC=CC2=N1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Appearance (Color)White
Melting Point (DSC)122.0-128.0?C
Assay (GC)≥98.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only