4-Bromo-2-fluorobenzoic acid, 98%
4-Bromo-2-fluorobenzoic acid, 98%
4-Bromo-2-fluorobenzoic acid, 98%
Thermo Scientific Chemicals

4-Bromo-2-fluorobenzoic acid, 98%

CAS: 112704-79-7 | C7H3BrFO2 | 218.00 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number B20118.06
also known as B20118-06
Price (USD)
79.65
Special offer
Online exclusive
Ends: 30-Jun-2026
94.10
Save 14.45 (15%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
79.65
Special offer
Online exclusive
Ends: 30-Jun-2026
94.10
Save 14.45 (15%)
Each
Chemical Identifiers
CAS112704-79-7
IUPAC Name4-bromo-2-fluorobenzoate
Molecular FormulaC7H3BrFO2
InChI KeyZQQSRVPOAHYHEL-UHFFFAOYSA-M
SMILES[O-]C(=O)C1=CC=C(Br)C=C1F
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
Melting Point (clear melt)209.0-215.0?C
Appearance (Color)White to pale cream
Assay (HPLC)≥97.5%
FormPowder
4-Bromo-2-fluorobenzoic acid may be used in the synthesis of biaryl intermediates by palladium-mediated coupling with various aryl boronic acids

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromo-2-fluorobenzoic acid may be used in the synthesis of biaryl intermediates by palladium-mediated coupling with various aryl boronic acids

Solubility
Solubility in methanol, very faint turbidity.

Notes
Store away from oxidizing agents, reducing agents and bases. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

General References:

  1. Jun Yong Choi, et al. R-Configuration of 4-Aminopyridyl-Based Inhibitors of CYP51 Confers Superior Efficacy Against Trypanosoma cruzi.Org. Biomol. Chem.,2003,1(8), 1256-1269.
  2. Fan Yang, et al. Oxidation of toluenes to benzoic acids by oxygen in non-acidic solvents.Tetrahedron.,200460(5), 1225-1228.