4-(1-Piperazinyl)phenol, 95%
4-(1-Piperazinyl)phenol, 95%
4-(1-Piperazinyl)phenol, 95%
4-(1-Piperazinyl)phenol, 95%
Thermo Scientific Chemicals

4-(1-Piperazinyl)phenol, 95%

CAS: 56621-48-8 | C10H14N2O | 178.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Catalog number B20122.18
also known as B20122-18
Price (USD)
189.65
Online Exclusive
211.00
Save 21.35 (10%)
Each
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)
189.65
Online Exclusive
211.00
Save 21.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS56621-48-8
IUPAC Name4-(piperazin-1-yl)phenol
Molecular FormulaC10H14N2O
InChI KeyGPEOAEVZTOQXLG-UHFFFAOYSA-N
SMILESOC1=CC=C(C=C1)N1CCNCC1
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Assay (Non-aqueous acid-base Titration)≥94.0 to ≤106.0%
Appearance (Color)Pale brown to brown or pink
Assay (HPLC)≥94.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only