Ethyl diacetoacetate, 97%
Ethyl diacetoacetate, 97%
Ethyl diacetoacetate, 97%
Thermo Scientific Chemicals

Ethyl diacetoacetate, 97%

CAS: 603-69-0 | C8H12O4 | 172.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number B20149.09
also known as B20149-09
Price (USD)
55.65
Online Exclusive
62.20
Save 6.55 (11%)
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
55.65
Online Exclusive
62.20
Save 6.55 (11%)
Each
Chemical Identifiers
CAS603-69-0
IUPAC Nameethyl 2-acetyl-3-oxobutanoate
Molecular FormulaC8H12O4
InChI KeyYMCDYRGMTRCAPZ-UHFFFAOYSA-N
SMILESCCOC(=O)C(C(C)=O)C(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥96.0%
Refractive Index1.4700-1.4750 @ 20?C
Appearance (Color)Clear colorless to yellow
FormLiquid
Ethyl diacetoacetate is used in the preparation of 3-acetyl-6-chloro-7-hydroxy-4-methylcoumarin. It is used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl diacetoacetate is used in the preparation of 3-acetyl-6-chloro-7-hydroxy-4-methylcoumarin. It is used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Not miscible in water.

Notes
Incompatible with oxidizing agents and bases. Store in a cool, dry conditions in a well sealed container.
RUO – Research Use Only
K F Chilvers; J D Perry; A L James; R H Reed. Synthesis and evaluation of novel fluorogenic substrates for the detection of bacterial beta-galactosidase. Journal of Applied Microbiology. 2001, 91, (6), 1118-1130.