1H,1H,2H,2H-Perfluorooctanol, 97%
1H,1H,2H,2H-Perfluorooctanol, 97%
1H,1H,2H,2H-Perfluorooctanol, 97%
1H,1H,2H,2H-Perfluorooctanol, 97%
Thermo Scientific Chemicals

1H,1H,2H,2H-Perfluorooctanol, 97%

CAS: 647-42-7 | C8H5F13O | 364.106 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Catalog number B20156.30
also known as B20156-30
Price (USD)
1,428.65
Online Exclusive
1,587.00
Save 158.35 (10%)
Each
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (USD)
1,428.65
Online Exclusive
1,587.00
Save 158.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS647-42-7
IUPAC Name3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctan-1-ol
Molecular FormulaC8H5F13O
InChI KeyGRJRKPMIRMSBNK-UHFFFAOYSA-N
SMILESOCCC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Identification (FTIR)Conforms
Appearance (Color)Clear colorless to pale yellow
FormLiquid
Assay (GC)≥96.0%
Refractive Index1.3135-1.3185 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1H,1H,2H,2H-Perfluorooctanol is used to prepare fluorinated aromatic diamine by reacting with 2-(4-nitrophenoxy)-4-nitrophenol through Mitsunobu reaction, which one reacts with dianhydrides such as 4,4′-(hexafluoroisopropylidene)diphthalic anhydride (6FDA), pyromellitic dianhydride (PMDA) and benzophenonetetracarboxylic dianhydride (BTDA) to prepare polyimides.

Solubility
Miscible with chloroform and methanol. Immiscible with water.

Notes
Incompatible with strong oxidizing agents.
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only

General References:

  1. Kim, N.; Murphy, M. C.; Soper, S. A.; Nikitopoulos, D. E. Liquid-liquid segmented flows in polycarbonate microchannels with cross-sectional expansions. Int. J. Multiphase Flow 2014, 58, 83-96.
  2. Yasani, B. R.; Martinelli, E.; Galli, G.; Glisenti, A.; Mieszkin, S.; Callow, M. E.; Callow, J. A. A comparison between different fouling-release elastomer coatings containing surface-active polymers. Biofouling 2014, 30 (4), 387-399.