Methyl 3-methylaminocrotonate, 97%
Methyl 3-methylaminocrotonate, 97%
Methyl 3-methylaminocrotonate, 97%
Thermo Scientific Chemicals

Methyl 3-methylaminocrotonate, 97%

CAS: 13412-12-9 | C6H11NO2 | 129.159 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number B20199.06
also known as B20199-06
Price (USD)
71.65
Special offer
Online exclusive
Ends: 30-Jun-2026
84.10
Save 12.45 (15%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
71.65
Special offer
Online exclusive
Ends: 30-Jun-2026
84.10
Save 12.45 (15%)
Each
Chemical Identifiers
CAS13412-12-9
IUPAC Namemethyl 3-(methylimino)butanoate
Molecular FormulaC6H11NO2
InChI KeyUPQBRLACXPZFPJ-UHFFFAOYSA-N
SMILESCOC(=O)CC(C)=NC
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Melting Point (clear melt)57.0-66.0?C
Assay (GC)≥96.0%
Appearance (Color)White to pale cream
It is employed as an intermediate in the synthesis of an analogue to amlodipine, bearing a short amino polyethylene glycol chain.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as an intermediate in the synthesis of an analogue to amlodipine, bearing a short amino polyethylene glycol chain.

Solubility
Soluble in water at (54 g/L) (25°C), Calc.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only
Jean Christophe Legeay, et al. Sequential synthesis of a new analogue of amlodipine bearing a short amino polyethyleneglycol chain.Tetrahedron.2007,63(48), 12081-12086.