3-Hexanol, 98%
3-Hexanol, 98%
3-Hexanol, 98%
Thermo Scientific™

3-Hexanol, 98%

CAS: 623-37-0 | C6H14O | 102.177 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number B20210.14
also known as B20210-14
Price (USD)
83.20
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
83.20
Each
Chemical Identifiers
CAS623-37-0
IUPAC Namehexan-3-ol
Molecular FormulaC6H14O
InChI KeyZOCHHNOQQHDWHG-UHFFFAOYNA-N
SMILESCCCC(O)CC
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥97.5%
Identification (FTIR)Conforms
Appearance (Color)Clear colorless
FormLiquid
Refractive Index1.4140-1.4180 @ 20?C
It finds it application as a flavoring agent or adjuvant.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It finds it application as a flavoring agent or adjuvant.

Solubility
Soluble in alcohol.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

General References:

  1. Ziółkowska NE, et al. New active HIV-1 protease inhibitors derived from 3-hexanol: conformation study of the free inhibitors in crystalline state and in complex with the enzyme.Chem Biol Drug Des.2012,79(5), 798-809.
  2. Jörg Friedrich.; Gerhard M Schneider. Near-infrared spectroscopic investigations on phase behaviour and association of 1-octadecanol, 3-hexanol, and 3-methyl-3-pentanol in carbon dioxide and chlorotrifluoromethane.The Journal of Chemical Thermodynamics.1989,21(3), 307-319.