Copper(II) trifluoromethanesulfonate, 98%
Copper(II) trifluoromethanesulfonate, 98%
Copper(II) trifluoromethanesulfonate, 98%
Thermo Scientific Chemicals

Copper(II) trifluoromethanesulfonate, 98%

CAS: 34946-82-2 | CHCuF3O3S | 213.617 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number B20253.06
also known as B20253-06
Price (USD)
79.65
Special offer
Online exclusive
Ends: 30-Jun-2026
93.90
Save 14.25 (15%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
79.65
Special offer
Online exclusive
Ends: 30-Jun-2026
93.90
Save 14.25 (15%)
Each
Chemical Identifiers
CAS34946-82-2
IUPAC Namecopper(II) triflate
Molecular FormulaC2H2CuF6O4S2
InChI KeyTXWAGEFXPDWAQH-UHFFFAOYSA-N
SMILES[Cu].FC(F)(F)S(=O)=O.FC(F)(F)S(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream to pale green or pale blue to pale gray
Assay (Iodometric Titration)≥97.5 to ≤102.5%
FormPowder
Copper(II) trifluoromethanesulfonate acts as a catalyst in Diels-Alder reaction and cyclopropanation reactions. It is also used as a reagent for oligosaccharide synthesis. It acts as a Lewis acid. It is also employed in the dimerization of ketone enolates and tetramethylsilane enol ethers. It is used to prepare cis vinylic sulfones and oxazoles from alkynyl sulfones and ketones respectively. Further, it plays an important role for cyclization of dienolates. In addition to this, it is useful for dehydration of alcohols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Copper(II) trifluoromethanesulfonate acts as a catalyst in Diels-Alder reaction and cyclopropanation reactions. It is also used as a reagent for oligosaccharide synthesis. It acts as a Lewis acid. It is also employed in the dimerization of ketone enolates and tetramethylsilane enol ethers. It is used to prepare cis vinylic sulfones and oxazoles from alkynyl sulfones and ketones respectively. Further, it plays an important role for cyclization of dienolates. In addition to this, it is useful for dehydration of alcohols.

Solubility
Soluble in water.

Notes
Hygroscopic. Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Mild Lewis acid:
  2. Used catalytically, promotes dehydration of alcohols and diols to alkenes at ambient temperatures: Helv. Chim. Acta, 70, 607 (1987).
  3. Widely used to generate carbenoid species from ɑ-diazo esters and ketones, via in situ reduction to the Cu(I) species: J. Am. Chem. Soc., 95, 3300 (1973). Preferred reagent for intramolecular cyclization of various diazo ketones: J. Org. Chem., 49, 11496 (1984). Also promotes the reaction between diazo esters and imines to give aziridines: J. Chem. Soc., Chem. Commun., 1 (1995).
  4. Catalyzes syn-selective aldol condensation of (Z)-silyl enol ethers with aldehydes: Chem. Lett., 959 (1997).
  5. Friedel-Crafts alkylation and acylation reactions of aromatics are also catalyzed: Tetrahedron, 57, 241 (2001).
  6. For a brief feature on uses of the reagent in synthesis, see: Synlett, 1044 (2005).
  7. Balaso, G. J.; Ashish, A. V.; Shriniwas, D. S. Copper(II)triflate Promoted Highly Chemoselective Rearrangement of Chalcone Epoxides to beta-keto Aldehydes. Lett. Org. Chem. 2015, 12 (1), 55-61.
  8. Jha, M.; Shelke, G. M.; Pericherla, K.; Kumar, A. Microwave assisted copper triflate-catalyzed rapid hydration of aryl acetylenes. Tetrahedron Lett. 2014, 55 (34), 4814-4816.