2-Ethyl-1,3-cyclopentanedione, 98%
2-Ethyl-1,3-cyclopentanedione, 98%
2-Ethyl-1,3-cyclopentanedione, 98%
Thermo Scientific Chemicals

2-Ethyl-1,3-cyclopentanedione, 98%

CAS: 823-36-9 | C7H10O2 | 126.155 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number B20254.06
also known as B20254-06
Price (USD)
36.65
Special offer
Online exclusive
Ends: 30-Jun-2026
42.40
Save 5.75 (14%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
36.65
Special offer
Online exclusive
Ends: 30-Jun-2026
42.40
Save 5.75 (14%)
Each
Chemical Identifiers
CAS823-36-9
IUPAC Name2-ethylcyclopentane-1,3-dione
Molecular FormulaC7H10O2
InChI KeyYDFBIBUYOUFJMR-UHFFFAOYSA-N
SMILESCCC1C(=O)CCC1=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
Melting Point (clear melt)168.0-178.0°C
Appearance (Color)White to cream to yellow to pale brown
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only