1-Hexene, 98%
1-Hexene, 98%
1-Hexene, 98%
Thermo Scientific Chemicals

1-Hexene, 98%

CAS: 592-41-6 | C6H12 | 84.162 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
Catalog number B20271.AP
also known as B20271-AP
Price (USD)
48.65
Online Exclusive
53.80
Save 5.15 (10%)
Each
Quantity:
500 mL
Request bulk or custom format
Price (USD)
48.65
Online Exclusive
53.80
Save 5.15 (10%)
Each
Chemical Identifiers
CAS592-41-6
IUPAC Namehex-1-ene
Molecular FormulaC6H12
InChI KeyLIKMAJRDDDTEIG-UHFFFAOYSA-N
SMILESCCCCC=C
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥97.5%
FormLiquid
Appearance (Color)Clear colorless
Identification (FTIR)Conforms
Refractive Index1.3855-1.3900 @ 20?C
1-Hexene is used as a solvent, paint thinner and a medium for conducting chemical reactions. It is a commoner and used in the preparation of both low density and high density polyethylenes. It acts as an adhesives, lubricants, lubricant additives and sealant chemicals. Further, it is used in the preparation of hex-1-en-3-one, heptanal and heptanoic acid. It is also employed in the synthesis of linear plasticizers, oxo-alcohols, motor fuels, automotive additives and biodegradable surfactants. In addition to this, it is used in the preparation of mercaptans, flavors and fragrances, alkyl metals, halides and alkyl silanes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Hexene is used as a solvent, paint thinner and a medium for conducting chemical reactions. It is a commoner and used in the preparation of both low density and high density polyethylenes. It acts as an adhesives, lubricants, lubricant additives and sealant chemicals. Further, it is used in the preparation of hex-1-en-3-one, heptanal and heptanoic acid. It is also employed in the synthesis of linear plasticizers, oxo-alcohols, motor fuels, automotive additives and biodegradable surfactants. In addition to this, it is used in the preparation of mercaptans, flavors and fragrances, alkyl metals, halides and alkyl silanes.

Solubility
Miscible with water, ethanol, ethyl ether, benzene and petroleum ether.

Notes
Store in a cool place. Incompatible with strong oxidizing agents and strong acids.
RUO – Research Use Only

General References:

  1. Nawdiyal, A.; Hansen, N.; Zeuch, T.; Seidel, L.; Mauss, F. Experimental and modelling study of speciation and benzene formation pathways in premixed 1-hexene flames. Proc. Combust. Inst. 2015, 35 (1), 325-332.
  2. Despagnet-Ayoub, E.; Takase, M. K.; Henling, L. M.; Labinger, J. A.; Bercaw, J. E. Mechanistic Insights on the Controlled Switch from Oligomerization to Polymerization of 1-Hexene Catalyzed by an NHC-Zirconium Complex. Organometallics 2015, 34 (19), 4707-4716.